Gene Transcripts
- Gene Symbol
- SNCAIP
- Entrez Gene ID
- 9627
- Gene Name
- synuclein alpha interacting protein
- Gene Aliases
- SYPH1, Sph1
- Chromosome Location
- Chr. 5: 122311354 - 122464219 on Build GRCh38
- UniGene ID
- Hs.426463
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308105.1 |
NP_001295034.1
|
8, 9 |
2629 |
|
NM_001308106.1 |
NP_001295035.1
|
8, 9 |
1898 |
|
NM_001308107.1 |
NP_001295036.1
|
8, 9 |
1973 |
|
NM_001308108.1 |
NP_001295037.1
|
8, 9 |
1937 |
|
NM_001308109.1 |
NP_001295038.1
|
7, 8 |
1859 |
|
NM_005460.3 |
NP_005451.2
|
10, 11 |
3025 |
|
NR_131761.1 |
|
12, 13 |
3175 |
|
NR_131762.1 |
|
9, 10 |
1904 |
|
XM_017010081.1 |
XP_016865570.1
|
11, 12 |
3081 |
|
XM_017010082.1 |
XP_016865571.1
|
10, 11 |
2912 |
|
XM_017010085.1 |
XP_016865574.1
|
9, 10 |
1968 |
|
GenBank mRNA |
AF076929.1 |
- |
10, 11 |
2856 |
|
AK021944.1 |
- |
3, 4 |
1748 |
|
AK299687.1 |
- |
9, 10 |
1904 |
|
AK304646.1 |
- |
7, 8 |
1691 |
|
BC033743.1 |
- |
8, 9 |
1793 |
5193089 |
View Full Product Details