Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004748.4 | NP_004739.3 | 8 | 2870 | |
NM_020739.3 | NP_065790.2 | 8 | 2630 | ||
GenBank mRNA | AB033080.2 | - | 8 | 2415 | |
AF011794.1 | - | 1 | 1197 | ||
AF212228.1 | - | 8 | 2540 | ||
BC034914.1 | - | 3 | 2788 | 5276323 | |
BC039871.1 | - | 8 | 2384 | 4385087 |