Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001307924.1 | NP_001294853.1 | 12 | 1491 | |
NM_004537.5 | NP_004528.1 | 12 | 1491 | ||
NM_139207.3 | NP_631946.1 | 12 | 1491 | ||
XM_006719425.3 | XP_006719488.1 | 10 | 954 | ||
XM_011538390.2 | XP_011536692.2 | 12 | 1150 | ||
XM_011538393.2 | XP_011536695.1 | 12 | 1403 | ||
XM_017019338.1 | XP_016874827.1 | 12 | 1403 | ||
XM_017019339.1 | XP_016874828.1 | 10 | 1214 | ||
XM_017019340.1 | XP_016874829.1 | 10 | 954 | ||
GenBank mRNA | AK122670.1 | - | 10 | 1025 | |
AK295147.1 | - | 12 | 1070 | ||
AK297500.1 | - | 9 | 963 | ||
AK304787.1 | - | 11 | 1104 | ||
AL162068.1 | - | 12 | 1128 | ||
BC002387.2 | - | 12 | 1125 | 2964631 | |
BC015599.1 | - | 11 | 1340 | ||
BT007023.1 | - | 11 | 1004 | ||
M86667.1 | - | 12 | 1079 |