Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190799.1 | NP_001177728.1 | 10 | 2425 | |
NM_001190801.1 | NP_001177730.1 | 11 | 2344 | ||
NM_014264.4 | NP_055079.3 | 11 | 2521 | ||
XM_005262701.2 | XP_005262758.1 | 11 | 2381 | ||
XM_017007662.1 | XP_016863151.1 | 11 | 2680 | ||
XM_017007663.1 | XP_016863152.1 | 11 | 2579 | ||
GenBank mRNA | AB006972.1 | - | 11 | 2435 | |
AK302858.1 | - | 11 | 2344 | ||
AK303399.1 | - | 10 | 2359 | ||
AK314238.1 | - | 11 | 2489 | ||
BC036023.1 | - | 11 | 2489 | 5273226 | |
Y13115.1 | - | 11 | 2387 |