Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013581.3 | NP_038609.3 | 5 | 1014 | |
GenBank mRNA | AF109377.2 | - | 5 | 997 | |
AK031510.1 | - | 4 | 470 | ||
AK034636.2 | - | 5 | 1019 | ||
AK152464.1 | - | 5 | 1005 | ||
AK158507.1 | - | 5 | 1376 | ||
AK159497.1 | - | 5 | 1016 | ||
AK162971.1 | - | 5 | 1015 | ||
AK171574.1 | - | 5 | 1008 | ||
AK207477.1 | - | 3 | 273 | ||
AK213132.1 | - | 3 | 319 | ||
AK220387.1 | - | 5 | 1011 | ||
BC063056.1 | - | 5 | 996 | 5720460 |