Gene Transcripts

Gene Symbol
Kif21b
Entrez Gene ID
16565
Gene Name
kinesin family member 21B
Gene Aliases
2610511N21Rik, mKIAA0449
Chromosome Location
Chr. 1: 136131374 - 136178014 on Build GRCm38
UniGene ID
Mm.276135
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039472.1 NP_001034561.1 2 512
GenBank mRNA AF202893.1 - 2 490
AK089400.1 - 2 460
BY742264.1 - 2 482

View Full Product Details