Gene Transcripts

Gene Symbol
Hus1
Entrez Gene ID
15574
Gene Name
Hus1 homolog (S. pombe)
Gene Aliases
Chromosome Location
Chr. 11: 8993137 - 9011534 on Build GRCm38
UniGene ID
Mm.42201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF076845.1 - 8 924
AK046152.1 - 8 931
AK051839.1 - 7 1356
AK088403.1 - 8 934
AK135922.1 - 8 914
BC047460.1 - 8 941

View Full Product Details