Gene Transcripts
- Gene Symbol
- Emp3
- Entrez Gene ID
- 13732
- Gene Name
- epithelial membrane protein 3
- Gene Aliases
- H-4, H4, HNMP-1, MI-35, Ymp
- Chromosome Location
- Chr. 7: 45918023 - 45921426 on Build GRCm38
- UniGene ID
- Mm.20829
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC001999.1 |
- |
5 |
531 |
|
U87948.1 |
- |
5 |
664 |
|
View Full Product Details