Gene Transcripts
- Gene Symbol
- En1
- Entrez Gene ID
- 13798
- Gene Name
- engrailed 1
- Gene Aliases
- En-1, Mo-en.1
- Chromosome Location
- Chr. 1: 120602391 - 120607991 on Build GRCm38
- UniGene ID
- Mm.2657
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L12703.1 |
- |
2 |
1749 |
|
View Full Product Details