Gene Transcripts

Gene Symbol
Bub3
Entrez Gene ID
12237
Gene Name
budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae)
Gene Aliases
AU019800, AU021329, AU043350, AW146323, C78067
Chromosome Location
Chr. 7: 131560370 - 131571898 on Build GRCm38
UniGene ID
Mm.441749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009774.3 NP_033904.2 8 1891
GenBank mRNA AK012744.1 - 3 1095
AK088534.1 - 8 1892
BC086790.1 - 6 3856

View Full Product Details