Gene Transcripts

Gene Symbol
Arpc1b
Entrez Gene ID
11867
Gene Name
actin related protein 2/3 complex, subunit 1B
Gene Aliases
41kDa, AA408064, AA408534, AA571392, AF007010, AW208418, L72, SOP2Hs, p41-ARC
Chromosome Location
Chr. 5: 145114256 - 145128186 on Build GRCm38
UniGene ID
Mm.30010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF162768.1 - 9 1228
AK148145.1 - 10 1297
AK206912.1 - 1 125
AK207221.1 - 1 125
BC003441.1 - 10 1235 2650641
BC010275.1 - 10 1245 3589768

View Full Product Details