Gene Transcripts
- Gene Symbol
- WDR46
- Entrez Gene ID
- 9277
- Gene Name
- WD repeat domain 46
- Gene Aliases
- BING4, C6orf11, FP221, UTP7
- Chromosome Location
- Chr. 6: 33278850 - 33289527 on Build GRCh38
- UniGene ID
- Hs.520063
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF447870.1 |
- |
15 |
2041 |
|
BC000388.2 |
- |
15 |
2036 |
2821198 |
View Full Product Details