Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037987.3 | NP_001033076.1 | 4 | 781 | |
NM_010103.4 | NP_034233.1 | 3 | 751 | ||
GenBank mRNA | AF031524.1 | - | 4 | 889 | |
AF031525.1 | - | 4 | 820 | ||
AK036117.1 | - | 4 | 742 | ||
AK044978.1 | - | 3 | 610 | ||
AK045088.1 | - | 3 | 567 | ||
AK046885.1 | - | 4 | 782 | ||
AK081003.1 | - | 3 | 583 | ||
AK084215.1 | - | 4 | 636 | ||
AK135220.1 | - | 4 | 745 | ||
AK139093.1 | - | 4 | 743 | ||
AK141907.1 | - | 4 | 765 | ||
AK157810.1 | - | 4 | 766 | ||
BC056386.1 | - | 4 | 681 | 6854497 |