Gene Transcripts

Gene Symbol
Magee1
Entrez Gene ID
107528
Gene Name
melanoma antigen, family E, 1
Gene Aliases
AI847422, DAMAGE, mMage-e1
Chromosome Location
Chr. X: 105120396 - 105123912 on Build GRCm38
UniGene ID
Mm.24341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053201.3 NP_444431.3 1 1650
GenBank mRNA AF319978.1 - 1 853
AK030032.1 - 1 1644
AK051625.1 - 1 1624
AK081378.1 - 1 1626
BC059039.1 - 1 1650 6812964

View Full Product Details