Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318872.1 | NP_001305801.1 | 2 | 652 | |
NM_006567.4 | NP_006558.1 | 2 | 646 | ||
XM_005248812.3 | XP_005248869.1 | 3 | 710 | ||
XM_006714966.2 | XP_006715029.1 | 2 | 621 | ||
XM_011514247.2 | XP_011512549.1 | 2 | 621 | ||
XM_011514248.2 | XP_011512550.1 | 2 | 621 | ||
XM_011514249.1 | XP_011512551.1 | 2 | 621 | ||
XM_011514251.2 | XP_011512553.1 | 2 | 621 | ||
XM_017010186.1 | XP_016865675.1 | 2 | 1429 | ||
XM_017010187.1 | XP_016865676.1 | 3 | 1431 | ||
GenBank mRNA | AF097441.1 | - | 1 | 302 | |
AK223423.1 | - | 2 | 446 | ||
AK312454.1 | - | 2 | 626 | ||
BC020239.1 | - | 2 | 446 | 5258249 | |
BC021112.1 | - | 2 | 446 | 5088776 | |
BE792999.1 | - | 2 | 591 | ||
BI761156.1 | - | 2 | 468 | ||
CN292346.1 | - | 2 | 480 | ||
CR542279.1 | - | 1 | 290 |