Gene Transcripts

Gene Symbol
Chchd5
Entrez Gene ID
66170
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
1110027L01Rik, AW045710
Chromosome Location
Chr. 2: 129129700 - 129134112 on Build GRCm38
UniGene ID
Mm.40621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025395.3 NP_079671.1 4 899
GenBank mRNA AK003936.1 - 4 781
AK013018.1 - 4 778
BC027303.1 - 2 458

View Full Product Details