Gene Transcripts

Gene Symbol
Gkn2
Entrez Gene ID
66284
Gene Name
gastrokine 2
Gene Aliases
1810036H07Rik, Bricd1b
Chromosome Location
Chr. 6: 87373365 - 87379494 on Build GRCm38
UniGene ID
Mm.46446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025467.1 NP_079743.1 3 161
GenBank mRNA AK007451.1 - 3 161
AK007705.1 - 3 161
AK008692.1 - 3 162
AK008986.1 - 3 162
AY943909.1 - 3 169
BC099445.1 - 3 150 30788014

View Full Product Details