Gene Transcripts

Gene Symbol
Bysl
Entrez Gene ID
53414
Gene Name
bystin-like
Gene Aliases
Bys, Enp1
Chromosome Location
Chr. 17: 47599331 - 47611492 on Build GRCm38
UniGene ID
Mm.472101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016859.3 NP_058555.3 7 2765
GenBank mRNA AK049195.1 - 7 2758
AK051704.1 - 7 2755
AK083033.1 - 7 2752
AK143027.1 - 7 2766

View Full Product Details