Gene Transcripts

Gene Symbol
Slc25a4
Entrez Gene ID
85333
Gene Name
solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4
Gene Aliases
Ant1
Chromosome Location
Chr. 16: 49266903 - 49270698 on Build Rnor_6.0
UniGene ID
Rn.4092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053515.1 NP_445967.1 2 629
GenBank mRNA BC060533.1 - 2 633
D12770.1 - 2 596
FQ213027.1 - 2 626
FQ213086.1 - 2 626
FQ214384.1 - 2 625
FQ215448.1 - 2 629
FQ217254.1 - 2 625
FQ220525.1 - 2 624
FQ224814.1 - 2 626

View Full Product Details