Gene Transcripts

Gene Symbol
Cxcr3
Entrez Gene ID
84475
Gene Name
chemokine (C-X-C motif) receptor 3
Gene Aliases
Gpr9
Chromosome Location
Chr. X: 71614346 - 71616997 on Build Rnor_6.0
UniGene ID
Rn.24787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053415.1 NP_445867.1 2 1034
GenBank mRNA AF223642.1 - 2 1034
BC091136.1 - 1 889 7376065

View Full Product Details