Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
287006
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
GST-2
Chromosome Location
Chr. 9: 104622219 - 104700657 on Build Rnor_6.0
UniGene ID
Rn.86443
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173338.1 NP_775460.1 169
GenBank mRNA AF326113.1 - 169

Gene Symbol
LOC102554041
Entrez Gene ID
102554041
Gene Name
solute carrier organic anion transporter family member 6A1-like
Gene Aliases
Chromosome Location
Chr. 9: 104716078 - 104719147 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767397.1 XP_008765619.1 1 338

View Full Product Details