Gene Transcripts

Gene Symbol
Cyp27a1
Entrez Gene ID
301517
Gene Name
cytochrome P450, family 27, subfamily a, polypeptide 1
Gene Aliases
Cyp27, P450C27
Chromosome Location
Chr. 9: 81968289 - 81998213 on Build Rnor_6.0
UniGene ID
Rn.94956
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245175.1 XP_006245237.1 4 696
GenBank mRNA BC061848.1 - 4 860
M38566.1 - 4 861

View Full Product Details