Gene Transcripts

Gene Symbol
Gucy2d
Entrez Gene ID
113911
Gene Name
guanylate cyclase 2d
Gene Aliases
Chromosome Location
Chr. 1: 163398891 - 163433594 on Build Rnor_6.0
UniGene ID
Rn.10067
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130737.4 NP_570093.1 7 1724
XM_006229710.1 XP_006229772.1 7 1723
GenBank mRNA L37203.1 - 7 1724

View Full Product Details