Gene Transcripts

Gene Symbol
Cyp7a1
Entrez Gene ID
25428
Gene Name
cytochrome P450, family 7, subfamily a, polypeptide 1
Gene Aliases
CHAP, CYP7, CYP7S1
Chromosome Location
Chr. 5: 19358734 - 19368431 on Build Rnor_6.0
UniGene ID
Rn.10737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012942.2 NP_037074.1 3 407
GenBank mRNA J05430.1 - 3 402
J05460.1 - 3 408
J05509.1 - 3 419

View Full Product Details