Gene Transcripts

Gene Symbol
LMOD1
Entrez Gene ID
25802
Gene Name
leiomodin 1
Gene Aliases
1D, 64kD, D1, SM-LMOD, SMLMOD
Chromosome Location
Chr. 1: 201896456 - 201946588 on Build GRCh38
UniGene ID
Hs.519075
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012134.2 NP_036266.2 3 3382
GenBank mRNA AK126474.1 - 3 2921
BC080187.1 - 3 3331 6177902
X54162.1 - 3 3262

View Full Product Details