Gene Transcripts
- Gene Symbol
- KIF4A
- Entrez Gene ID
- 24137
- Gene Name
- kinesin family member 4A
- Gene Aliases
- KIF4, KIF4G1, MRX100
- Chromosome Location
- Chr. X: 70290029 - 70420924 on Build GRCh38
- UniGene ID
- Hs.648326
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012310.4 |
NP_036442.3
|
7, 8 |
928 |
|
GenBank mRNA |
AB208797.1 |
- |
7, 8 |
828 |
|
AF071592.2 |
- |
7, 8 |
845 |
|
AF179308.1 |
- |
6, 7 |
785 |
|
AF277375.1 |
- |
4, 5 |
404 |
|
AJ271784.1 |
- |
6, 7 |
791 |
|
AK022592.1 |
- |
4, 5 |
388 |
|
AK313133.1 |
- |
6, 7 |
1161 |
|
BC003664.2 |
- |
7, 8 |
866 |
|
BC038459.1 |
- |
7, 8 |
865 |
|
BC049218.1 |
- |
7, 8 |
834 |
|
BC050548.1 |
- |
7, 8 |
827 |
4538604 |
- Gene Symbol
- KIF4B
- Entrez Gene ID
- 285643
- Gene Name
- kinesin family member 4B
- Gene Aliases
- Chromosome Location
- Chr. 5: 155013700 - 155018125 on Build GRCh38
- UniGene ID
- Hs.567824
- Species
- Homo sapiens
View Full Product Details