Gene Transcripts
- Gene Symbol
- C18orf8
- Entrez Gene ID
- 29919
- Gene Name
- chromosome 18 open reading frame 8
- Gene Aliases
- HsT2591, MIC1, Mic-1
- Chromosome Location
- Chr. 18: 23503470 - 23531807 on Build GRCh38
- UniGene ID
- Hs.464779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276342.1 |
NP_001263271.1
|
1, 2 |
259 |
|
NM_001318707.1 |
NP_001305636.1
|
1, 2 |
259 |
|
NM_001318708.1 |
NP_001305637.1
|
1, 2 |
259 |
|
NM_001318709.1 |
NP_001305638.1
|
1, 2 |
259 |
|
NM_013326.4 |
NP_037458.3
|
1, 2 |
259 |
|
XM_011525939.1 |
XP_011524241.1
|
1, 2 |
216 |
|
XM_017025717.1 |
XP_016881206.1
|
1, 2 |
210 |
|
GenBank mRNA |
AK057192.1 |
- |
1, 2 |
220 |
|
AK295246.1 |
- |
1, 2 |
216 |
|
AK296230.1 |
- |
1, 2 |
210 |
|
BC002950.1 |
- |
1, 2 |
184 |
3538017 |
BC008305.1 |
- |
1, 2 |
191 |
3537084 |
CN481504.1 |
- |
1, 2 |
231 |
|
DC383182.1 |
- |
1, 2 |
243 |
|
View Full Product Details