Gene Transcripts

Gene Symbol
LINC00312
Entrez Gene ID
29931
Gene Name
long intergenic non-protein coding RNA 312
Gene Aliases
ERR-10, ERR10, LMCD1DN, LOH3CR2A, NAG-7, NAG7, NCRNA00312
Chromosome Location
Chr. 3: 8571782 - 8574668 on Build GRCh38
UniGene ID
Hs.433151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024065.2 1 1226
GenBank mRNA AF086709.2 - 1 803
AK054898.1 - 1 1226
BC016278.1 - 1 924

View Full Product Details