Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099.4 | NP_001090.2 | 2 | 275 | |
NM_001134194.1 | NP_001127666.1 | 2 | 275 | ||
NM_001292037.1 | NP_001278966.1 | 2 | 275 | ||
XM_011512946.1 | XP_011511248.1 | 2 | 239 | ||
XM_011512947.2 | XP_011511249.1 | 2 | 239 | ||
GenBank mRNA | AB102888.1 | - | 2 | 185 | |
AK300540.1 | - | 2 | 236 | ||
BC007460.1 | - | 2 | 235 | 3951204 | |
BC008493.1 | - | 2 | 232 | 4272289 | |
BC016344.1 | - | 2 | 239 | 3950907 | |
BC017877.1 | - | 2 | 225 | ||
DC383453.1 | - | 2 | 275 | ||
M24902.1 | - | 2 | 208 | ||
M34840.1 | - | 2 | 199 | ||
X52174.1 | - | 2 | 208 | ||
X53605.1 | - | 2 | 221 |