Gene Transcripts

Gene Symbol
EHD3
Entrez Gene ID
30845
Gene Name
EH domain containing 3
Gene Aliases
PAST3
Chromosome Location
Chr. 2: 31234014 - 31269451 on Build GRCh38
UniGene ID
Hs.368808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014600.2 NP_055415.1 4 1438
XM_011532806.2 XP_011531108.1 2 402
GenBank mRNA AF181264.1 - 4 874
AF214736.1 - 4 1115

View Full Product Details