Gene Transcripts

Gene Symbol
KLHDC7A
Entrez Gene ID
127707
Gene Name
kelch domain containing 7A
Gene Aliases
Chromosome Location
Chr. 1: 18480930 - 18485999 on Build GRCh38
UniGene ID
Hs.406913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152375.2 NP_689588.2 1 1428
GenBank mRNA AK096072.1 - 1 986
BC101498.1 - 1 794
BC101500.1 - 1 794
BC151205.1 - 1 1187

View Full Product Details