Gene Transcripts

Gene Symbol
OR51F1
Entrez Gene ID
256892
Gene Name
olfactory receptor family 51 subfamily F member 1 (gene/pseudogene)
Gene Aliases
OR11-21, OR51F1P
Chromosome Location
Chr. 11: 4768979 - 4769917 on Build GRCh38
UniGene ID
Hs.553659
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004752.1 NP_001004752.1 1 465

View Full Product Details