Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001934.1 | NP_001001934.1 | 3 | 452 | |
XM_008772747.1 | XP_008770969.1 | 4 | 1139 | ||
GenBank mRNA | HE864428.1 | - | 2 | 363 | |
HE864429.1 | - | 3 | 492 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
GenBank mRNA | HE864431.1 | - | 735 | ||
HE864432.1 | - | 1884 | |||
HE864433.1 | - | 2109 | |||
HE864434.1 | - | 2365 |