Gene Transcripts

Gene Symbol
Mpi
Entrez Gene ID
300741
Gene Name
mannose phosphate isomerase (mapped)
Gene Aliases
Chromosome Location
Chr. 8: 62324176 - 62332080 on Build Rnor_6.0
UniGene ID
Rn.229358
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004081.1 NP_001004081.1 6 792
GenBank mRNA BC079111.1 - 6 792

View Full Product Details