Gene Transcripts

Gene Symbol
Icmt
Entrez Gene ID
170818
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
fcmt
Chromosome Location
Chr. 5: 169475270 - 169482031 on Build Rnor_6.0
UniGene ID
Rn.48709
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133310.2 NP_579844.2 5 964
GenBank mRNA AF075595.1 - 5 752
BC083923.1 - 6 1169

View Full Product Details