Gene Transcripts

Gene Symbol
TECPR2
Entrez Gene ID
9895
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
KIAA0329, SPG49
Chromosome Location
Chr. 14: 102362912 - 102502481 on Build GRCh38
UniGene ID
Hs.195667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014844.4 NP_055659.2 20 6075
GenBank mRNA AB002295.1 - 17 5378
AB002327.1 - 20 6024
AK127721.1 - 10 3503
BC136647.1 - 19 5872 9020637
BC151230.1 - 20 6048 8860116

View Full Product Details