Gene Transcripts

Gene Symbol
Poc1b
Entrez Gene ID
382406
Gene Name
POC1 centriolar protein homolog B (Chlamydomonas)
Gene Aliases
4933430F16Rik, Wdr51b
Chromosome Location
Chr. 10: 99107171 - 99198074 on Build GRCm38
UniGene ID
Mm.440656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027740.6 NP_082016.1 3 165
GenBank mRNA AK016991.1 - 2 89
AK030180.1 - 2 59
AK045302.1 - 3 144
AK050233.1 - 3 234
AK090075.1 - 3 168
BC050888.1 - 2 163
BC099926.1 - 3 252
BC144819.1 - 3 254
BC145801.1 - 3 254 40131104

View Full Product Details