Gene Transcripts
- Gene Symbol
- CYP2C9
- Entrez Gene ID
- 1559
- Gene Name
- cytochrome P450 family 2 subfamily C member 9
- Gene Aliases
- CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
- Chromosome Location
- Chr. 10: 94905867 - 94989395 on Build GRCh38
- UniGene ID
- Hs.282624
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000771.3 |
NP_000762.2
|
5, 6 |
852 |
|
XM_017015758.1 |
XP_016871247.1
|
6, 7 |
1272 |
|
GenBank mRNA |
AK289420.1 |
- |
5, 6 |
852 |
|
AK298458.1 |
- |
5, 6 |
852 |
|
BC125054.1 |
- |
5, 6 |
827 |
40116637 |
D00173.1 |
- |
5, 6 |
818 |
|
M15331.1 |
- |
5, 6 |
812 |
|
M21939.1 |
- |
5, 6 |
813 |
|
M21940.1 |
- |
3, 4 |
506 |
|
M61855.1 |
- |
5, 6 |
839 |
|
M61857.1 |
- |
5, 6 |
837 |
|
S46963.1 |
- |
5, 6 |
790 |
|
View Full Product Details