Gene Transcripts

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
Chromosome Location
Chr. 11: 74696428 - 74731385 on Build GRCh38
UniGene ID
Hs.432379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278473.2 NP_001265402.1 7, 8 1240
NM_001304390.1 NP_001291319.1 6, 7 1132
NM_001304416.1 NP_001291345.1 5, 6 1038
NM_001304417.1 NP_001291346.1 4, 5 830
NM_015424.5 NP_056239.3 7, 8 1240
GenBank mRNA AF332891.1 - 4, 5 521
AK304519.1 - 7, 8 1008
AL110168.1 - 1, 2 142
AY163868.1 - 7, 8 746
AY279090.1 - 7, 8 1037
AY279092.1 - 5, 6 1038
AY279093.1 - 5, 6 1038
AY279094.1 - 4, 5 944
AY358522.1 - 7, 8 1029
BC142623.1 - 7, 8 1240 40147467

View Full Product Details