Gene Transcripts

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
Chromosome Location
Chr. 22: 42508335 - 42519823 on Build GRCh38
UniGene ID
Hs.745006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015703.4 NP_056518.2 5 579
GenBank mRNA AK092492.1 - 3 1105
AK314650.1 - 5 545
AY039240.1 - 4 451
BC031838.1 - 4 494
BC035992.1 - 5 558
BC041639.1 - 5 537
BC042335.1 - 5 540
BC063537.1 - 1 75
BC073834.1 - 5 552 5533064
BC121118.1 - 5 562 40120513
BC121119.1 - 5 562 40120514

Gene Symbol
RRP7BP
Entrez Gene ID
91695
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
Gene Aliases
RRP7B, dJ222E13.2
Chromosome Location
Chr. 22: 42573244 - 42582011 on Build GRCh38
UniGene ID
Hs.534041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002184.2 5 545
GenBank mRNA AK074492.1 - 3 306
AK098201.1 - 5 572
AL157851.1 - 5 530
BC007736.2 - 5 545
BC014647.1 - 5 543
BC143340.1 - 5 633

View Full Product Details