Gene Transcripts

Gene Symbol
LRRTM3
Entrez Gene ID
347731
Gene Name
leucine rich repeat transmembrane neuronal 3
Gene Aliases
Chromosome Location
Chr. 10: 66926034 - 67101551 on Build GRCh38
UniGene ID
Hs.652155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178011.4 NP_821079.3 3 2575
NR_111909.1 3 619
GenBank mRNA AK294029.1 - 2 795
BC111492.1 - 3 2527 5196989
BX640611.1 - 3 2553

View Full Product Details