Gene Transcripts
- Gene Symbol
- REEP2
- Entrez Gene ID
- 51308
- Gene Name
- receptor accessory protein 2
- Gene Aliases
- C5orf19, SGC32445, SPG72, Yip2d
- Chromosome Location
- Chr. 5: 138439001 - 138446969 on Build GRCh38
- UniGene ID
- Hs.416090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271803.1 |
NP_001258732.1
|
4, 5 |
513 |
|
NM_016606.3 |
NP_057690.2
|
4, 5 |
513 |
|
NR_073448.1 |
|
4, 5 |
588 |
|
NR_073449.1 |
|
4, 5 |
588 |
|
GenBank mRNA |
AF251041.1 |
- |
3, 4 |
286 |
|
AK056193.1 |
- |
4, 5 |
469 |
|
AK223611.1 |
- |
4, 5 |
454 |
|
AK290547.1 |
- |
4, 5 |
403 |
|
AK291101.1 |
- |
4, 5 |
466 |
|
AK293480.1 |
- |
4, 5 |
487 |
|
AY562240.1 |
- |
4, 5 |
305 |
|
BC006218.2 |
- |
4, 5 |
427 |
3940709 |
DC424086.1 |
- |
4, 5 |
539 |
|
View Full Product Details