Gene Transcripts

Gene Symbol
Best1
Entrez Gene ID
293735
Gene Name
bestrophin 1
Gene Aliases
Vmd2
Chromosome Location
Chr. 1: 226033093 - 226049896 on Build Rnor_6.0
UniGene ID
Rn.136565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011940.1 NP_001011940.1 11 1645
XM_006231029.2 XP_006231091.1 11 1685
GenBank mRNA BC079048.1 - 11 1645

View Full Product Details