Gene Transcripts

Gene Symbol
Tti2
Entrez Gene ID
290811
Gene Name
TELO2 interacting protein 2
Gene Aliases
RGD1310414
Chromosome Location
Chr. 16: 64737342 - 64745207 on Build Rnor_6.0
UniGene ID
Rn.12987
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013883.1 NP_001013905.1 1 457
GenBank mRNA BC082008.1 - 1 457

View Full Product Details