Gene Transcripts

Gene Symbol
QPCTL
Entrez Gene ID
54814
Gene Name
glutaminyl-peptide cyclotransferase like
Gene Aliases
gQC
Chromosome Location
Chr. 19: 45692402 - 45703990 on Build GRCh38
UniGene ID
Hs.631556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163377.1 NP_001156849.1 3, 4 727
NM_017659.3 NP_060129.2 4, 5 1009
GenBank mRNA AK000091.1 - 4, 5 818
AK172764.1 - 4, 5 817
AK222636.1 - 4, 5 802
BC011553.1 - 3, 4 513 4564051

View Full Product Details