Gene Transcripts

Gene Symbol
MYADML
Entrez Gene ID
151325
Gene Name
myeloid associated differentiation marker like (pseudogene)
Gene Aliases
Chromosome Location
Chr. 2: 33726061 - 33728217 on Build GRCh38
UniGene ID
Hs.528847
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003143.2 1 1457
GenBank mRNA AK057470.1 - 1 1444
BC031088.1 - 1 1447
BC038980.1 - 1 1414

View Full Product Details