Gene Transcripts

Gene Symbol
Tex24
Entrez Gene ID
541463
Gene Name
testis expressed gene 24
Gene Aliases
1700108N07Rik, TESF-1
Chromosome Location
Chr. 8: 27344323 - 27349188 on Build GRCm38
UniGene ID
Mm.84066
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013609.2 NP_001013627.1 3 1365
GenBank mRNA AK007143.1 - 1 223
AY552600.1 - 3 1365

View Full Product Details