Gene Transcripts

Gene Symbol
Hnrnpa3
Entrez Gene ID
229279
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3
Chromosome Location
Chr. 2: 75659230 - 75669407 on Build GRCm38
UniGene ID
Mm.316306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053263.1 NP_444493.1 6 667
NM_146130.3 NP_666242.2 6 733
NM_198090.2 NP_932758.1 6 733
GenBank mRNA AB201711.1 - 6 713
AF463524.1 - 6 645
AK133860.1 - 6 732
AK169663.1 - 6 715
BC023828.1 - 6 686 5343619
BC023908.1 - 6 702 5322033
BC024454.1 - 3 250
BC038364.1 - 6 715 5362605
BC057655.1 - 6 646
BC062198.1 - 6 724 2655879
BC064824.1 - 6 698 6477672
BC085474.1 - 5 608 30538435
BC092083.1 - 3 328
BC116250.1 - 6 636 40093600
BC116251.1 - 6 636 40093602
BC157919.1 - 6 636 9007347
BC158038.1 - 6 636 9007341
CO045618.1 - 5 517

Gene Symbol
Gm6793
Entrez Gene ID
627828
Gene Name
heterogeneous nuclear ribonucleoprotein A3 pseudogene
Gene Aliases
EG627828
Chromosome Location
Chr. 8: 112013433 - 112015036 on Build GRCm38
UniGene ID
Mm.459502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC157904.1 - 1 636
BC158074.1 - 1 636 9007371
BC172011.1 - 1 636 9054297

View Full Product Details