Gene Transcripts

Gene Symbol
Supt7l
Entrez Gene ID
72195
Gene Name
suppressor of Ty 7-like
Gene Aliases
2610524B01Rik, 6030455L14Rik
Chromosome Location
Chr. 5: 31514569 - 31526762 on Build GRCm38
UniGene ID
Mm.164187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028150.2 NP_082426.1 5 2100
GenBank mRNA AK012134.1 - 5 2046
AK144989.1 - 5 1972
BC031447.1 - 5 2023 4502314

View Full Product Details