Gene Transcripts

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
Chromosome Location
Chr. 14: 93237550 - 93333092 on Build GRCh38
UniGene ID
Hs.525549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002860.3 NP_001002860.2 3 705
NM_018167.4 NP_060637.1 3 705
XM_011536939.2 XP_011535241.1 3 470
GenBank mRNA AB040958.2 - 3 634
AK001510.1 - 3 631
BC047071.1 - 3 702

View Full Product Details